Learn More
Benzenesulfonic acid, 94%
CAS: 98-11-3 | C6H6O3S | 158.171 g/mol
Supplier: Thermo Scientific Chemicals A161480E
Description
Benzenesulfonic acid is mainly consumed by conversion to other specialty chemicals. A variety of pharmaceutical drugs are prepared as salts of benzenesulfonic acid and are known as besylates or besilates. The alkali metal salt of benzenesulfonic acid was once widely used in the production of phenol. It acts as an acid catalyst for direct esterification of amino acids and peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzenesulfonic acid is mainly consumed by conversion to other specialty chemicals. A variety of pharmaceutical drugs are prepared as salts of benzenesulfonic acid and are known as besylates or besilates. The alkali metal salt of benzenesulfonic acid was once widely used in the production of phenol. It acts as an acid catalyst for direct esterification of amino acids and peptides.
Solubility
Soluble in water and alcohol. Slightly soluble in benzene. Insoluble in non-polar solvents.
Notes
It is hygroscopic. Incompatible with oxidizing agents and bases.
Specifications
| Benzenesulfonic acid | |
| ∼45°C to 50°C | |
| >110°C (230°F) | |
| MFCD00011689 | |
| UN2585 | |
| Hygroscopic | |
| benzenesulphonic acid, phenylsulfonic acid, besylic acid, benzene sulphonic acid, benzenemonosulfonic acid, benzene sulfonic acid, kyselina benzensulfonova, benzensulfonic acid, ccris 4595, kyselina benzensulfonova czech | |
| SRSXLGNVWSONIS-UHFFFAOYSA-N | |
| benzenesulfonic acid | |
| 7371 | |
| 158.18 |
| 98-11-3 | |
| 1.32 | |
| C6H6O3S | |
| 2500 g | |
| 742513 | |
| 14,1070 | |
| Soluble in water and alcohol. Slightly soluble in benzene. Insoluble in non-polar solvents. | |
| C1=CC=C(C=C1)S(=O)(=O)O | |
| 158.171 | |
| CHEBI:64455 | |
| 94% |
Chemical Identifiers
| 98-11-3 | |
| 158.171 | |
| SRSXLGNVWSONIS-UHFFFAOYSA-N | |
| 7371 | |
| benzenesulfonic acid |
| C6H6O3S | |
| MFCD00011689 | |
| benzenesulphonic acid, phenylsulfonic acid, besylic acid, benzene sulphonic acid, benzenemonosulfonic acid, benzene sulfonic acid, kyselina benzensulfonova, benzensulfonic acid, ccris 4595, kyselina benzensulfonova czech | |
| CHEBI:64455 | |
| C1=CC=C(C=C1)S(=O)(=O)O |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
EINECSNumber : 202-638-7
RTECSNumber : DB4200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only