Learn More
Benzenesulfinic acid sodium salt, 98%
CAS: 873-55-2 | C6H5NaO2S | 164.154 g/mol
Supplier: Thermo Scientific Chemicals A1211822
Description
It is used as a polymer adhesive enhancers, plasticizers for plasticized and modified of polyamide, epoxy resin, phenolic resin. It is designed for manufacturing of photography, pharmaceutical, electroplating industry, can be used as photo-reducing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a polymer adhesive enhancers, plasticizers for plasticized and modified of polyamide, epoxy resin, phenolic resin. It is designed for manufacturing of photography, pharmaceutical, electroplating industry, can be used as photo-reducing agent.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidants. Store in cool, dry place in a tightly sealed container.
Specifications
| Benzenesulfinic acid sodium salt | |
| >300°C | |
| MFCD00013135 | |
| 3598405 | |
| Soluble in water. | |
| C1=CC=C(C=C1)S(=O)[O-].[Na+] | |
| 164.154 | |
| 164.16 |
| 873-55-2 | |
| C6H5NaO2S | |
| 100 g | |
| sodium benzenesulfinate, benzenesulfinic acid sodium salt, benzenesulfinic acid, sodium salt, sodium phenylsulfinate, sodium benzenesulphinate, sodium benzene sulfinate, sodiumphenylsulfinate, phso2na, benzenesulfinic acid, sodium salt 1:1, sodium phenylsulphinate | |
| CHLCPTJLUJHDBO-UHFFFAOYSA-M | |
| sodium;benzenesulfinate | |
| 2723840 | |
| 98% |
Chemical Identifiers
| 873-55-2 | |
| 164.154 | |
| CHLCPTJLUJHDBO-UHFFFAOYSA-M | |
| 2723840 | |
| C1=CC=C(C=C1)S(=O)[O-].[Na+] |
| C6H5NaO2S | |
| MFCD00013135 | |
| sodium benzenesulfinate, benzenesulfinic acid sodium salt, benzenesulfinic acid, sodium salt, sodium phenylsulfinate, sodium benzenesulphinate, sodium benzene sulfinate, sodiumphenylsulfinate, phso2na, benzenesulfinic acid, sodium salt 1:1, sodium phenylsulphinate | |
| sodium;benzenesulfinate |
Safety and Handling
EINECSNumber : 212-842-8
RTECSNumber : DA9135000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only