missing translation for 'onlineSavingsMsg'
Learn More

Benzeneseleninic Acid 97.0+%, TCI America™

Catalog No. B21345G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B21345G Supplier TCI America Supplier No. B21345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6996-92-5
Molecular Formula C6H6O2Se
Molecular Weight (g/mol) 189.08
MDL Number MFCD00002100
InChI Key WIHKGDVGLJJAMC-UHFFFAOYSA-N
Synonym phenyl-selensaeure, benzene selenoic acid, seleninobenzoic acid, acmc-1bhr1, benzeneselenine acid, benzeneseleninic acid, chembl39469, benzeneseleninic acid, purum t
PubChem CID 23427
IUPAC Name benzeneseleninic acid
SMILES O[Se](=O)C1=CC=CC=C1

Specifications

Melting Point 122°C
Quantity 5 g
UN Number 3283
Formula Weight 189.07
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzeneseleninic Acid