missing translation for 'onlineSavingsMsg'
Learn More

Benzeneruthenium(II) Chloride Dimer 95.0+%, TCI America™

Catalog No. B19025G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19025G 5 g
B19021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19025G Supplier TCI America Supplier No. B19025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37366-09-9
Molecular Formula C12H12Cl4Ru2
Molecular Weight (g/mol) 500.168
MDL Number MFCD00064686
InChI Key YGXMUPKIEHNBNQ-UHFFFAOYSA-J
Synonym benzeneruthenium ii chloride dimer, dibenzenetetrachlorodiruthenium, bis benzene dichlororuthenium, benzene dichlororuthenium dimer, benzeneruthenium dichloride dimer, bis benzene tetrachlorodiruthenium, rucl2 benzene 2, dichloro benzene rutheniumdimer, benzene ruthenium cento chloride
PubChem CID 10962144
IUPAC Name benzene;dichlororuthenium
SMILES C1=CC=CC=C1.C1=CC=CC=C1.Cl[Ru]Cl.Cl[Ru]Cl

Specifications

Quantity 5 g
Formula Weight 500.17
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzeneruthenium(II) Chloride Dimer