missing translation for 'onlineSavingsMsg'
Learn More

Benzeneruthenium(II) chloride, dimer, 97%

CAS: 37366-09-9 | C12H12Cl4Ru2 | 500.18 g/mol

$309.18 - $309.18

Chemical Identifiers

CAS 37366-09-9
Molecular Formula C12H12Cl4Ru2
Molecular Weight (g/mol) 500.18
MDL Number MFCD00064686
InChI Key YGXMUPKIEHNBNQ-UHFFFAOYSA-J
Synonym benzeneruthenium ii chloride dimer, dibenzenetetrachlorodiruthenium, bis benzene dichlororuthenium, benzene dichlororuthenium dimer, benzeneruthenium dichloride dimer, bis benzene tetrachlorodiruthenium, rucl2 benzene 2, dichloro benzene rutheniumdimer, benzene ruthenium cento chloride
PubChem CID 10962144
IUPAC Name benzene;dichlororuthenium
SMILES C1=CC=CC=C1.C1=CC=CC=C1.Cl[Ru]Cl.Cl[Ru]Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC367480010
SDS View Documents
Thermo Scientific Chemicals
367480010
1 g Glass bottle
Each for $309.18
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

37366-09-9
500.18
YGXMUPKIEHNBNQ-UHFFFAOYSA-J
10962144
C1=CC=CC=C1.C1=CC=CC=C1.Cl[Ru]Cl.Cl[Ru]Cl
C12H12Cl4Ru2
MFCD00064686
benzeneruthenium ii chloride dimer, dibenzenetetrachlorodiruthenium, bis benzene dichlororuthenium, benzene dichlororuthenium dimer, benzeneruthenium dichloride dimer, bis benzene tetrachlorodiruthenium, rucl2 benzene 2, dichloro benzene rutheniumdimer, benzene ruthenium cento chloride
benzene;dichlororuthenium

Specifications

37366-09-9
100.0
Gray
97%
C12H12Cl4Ru2
1 g
benzeneruthenium ii chloride dimer, dibenzenetetrachlorodiruthenium, bis benzene dichlororuthenium, benzene dichlororuthenium dimer, benzeneruthenium dichloride dimer, bis benzene tetrachlorodiruthenium, rucl2 benzene 2, dichloro benzene rutheniumdimer, benzene ruthenium cento chloride
YGXMUPKIEHNBNQ-UHFFFAOYSA-J
benzene;dichlororuthenium
10962144
97%
Benzeneruthenium(II) chloride, dimer
96.0
249.0°C
Authentic
Glass bottle
MFCD00064686
07,109; 09,167; 11,182; 14,136; 15,135; 16,130
Solubility in water: insoluble
C1=CC=CC=C1.C1=CC=CC=C1.Cl[Ru]Cl.Cl[Ru]Cl
500.18
500.18
Turnings
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only