missing translation for 'onlineSavingsMsg'
Learn More

Benzamidoxime 98.0+%, TCI America™

Catalog No. B001525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B001525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B001525G Supplier TCI America Supplier No. B001525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 613-92-3
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00031485 MFCD00474011
InChI Key MXOQNVMDKHLYCZ-UHFFFAOYSA-N
Synonym benzamidoxime, benzamide oxime, n'-hydroxybenzimidamide, benzohydroxamamide, n-hydroxybenzamidine, phenylhydroxamidine, n-hydroxybenzenecarboximidamide, ccris 2953, n'-hydroxybenzamidine, benzamidoxim
PubChem CID 7259353
ChEBI CHEBI:83354
IUPAC Name (Z)-N'-hydroxybenzenecarboximidamide
SMILES N\C(=N/O)C1=CC=CC=C1

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 25 g
Formula Weight 136.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzamidoxime
RTECSNumber CY6920000
TSCA No