missing translation for 'onlineSavingsMsg'
Learn More

Benzamidine Hydrochloride 96.0+%, TCI America™

Catalog No. B001325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B001325G 25 g
B0013100G 100 g
B0013500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B001325G Supplier TCI America Supplier No. B001325G

Chemical Identifiers

CAS 1670-14-0
Molecular Formula C7H9ClN2
Molecular Weight (g/mol) 156.613
MDL Number MFCD00013025
InChI Key LZCZIHQBSCVGRD-UHFFFAOYSA-N
Synonym benzamidine hydrochloride, benzimidamide hydrochloride, benzenecarboximidamide hydrochloride, benzenecarboximidamide, monohydrochloride, benzamidinium chloride, benzamidine hcl, benzamidine, hydrochloride, amidinobenzene hydrochloride, benzamidine, monohydrochloride, amidinobenzene hydrochloride 1:1
PubChem CID 80289
IUPAC Name benzenecarboximidamide;hydrochloride
SMILES C1=CC=C(C=C1)C(=N)N.Cl

Specifications

Melting Point 171°C
Color White
pH 4.0 to 6.5 (50g/L 25°C)
Quantity 25 g
Formula Weight 156.61
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzamidine Hydrochloride
RTECSNumber CV6260000
TSCA Yes