missing translation for 'onlineSavingsMsg'
Learn More

Benzamidine hydrochloride hydrate, 99%, water ca 10-14%

Catalog No. p-7042524
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6282306 5 g
AAJ6282309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6282306 Supplier Thermo Scientific Chemicals Supplier No. J6282306
Only null left

Reversible inhibitor of trypsin, trypsin-like proteins and serine proteases | CAS: 206752-36-5 | C7H9ClN2 | 156.61 g/mol

Reversible inhibitor of trypsin, trypsin-like proteins and serine proteases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 206752-36-5
Molecular Formula C7H9ClN2
Molecular Weight (g/mol) 156.61
MDL Number MFCD00066285
InChI Key LZCZIHQBSCVGRD-UHFFFAOYSA-N
Synonym benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
PubChem CID 16219042
IUPAC Name benzenecarboximidamide;hydrate;hydrochloride
SMILES Cl.NC(=N)C1=CC=CC=C1

Specifications

Melting Point 80°C to 85°C
Color White
Quantity 5 g
Beilstein 3594959
Formula Weight 156.62
Percent Purity 99%
Physical Form Beads or Powder
Assay water ≈10 to 14%
Chemical Name or Material Benzamidine hydrochloride hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-795-4
RTECSNumber CV6260000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only