Learn More
Benzamidine hydrochloride hydrate, 98%, water ca 10-14%
CAS: 206752-36-5 | C7H9ClN2 | 156.61 g/mol
Supplier: Thermo Scientific Chemicals A1600214
Description
Benzamidine hydrochloride hydrate is a reversible inhibitor of trypsin, trypsin-like enzymes, and serine proteases. It is a proteolytic inhibitor and apoptosis reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzamidine hydrochloride hydrate is a reversible inhibitor of trypsin, trypsin-like enzymes, and serine proteases. It is a proteolytic inhibitor and apoptosis reagent.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
Specifications
| Benzamidine hydrochloride hydrate | |
| 82°C to 85°C | |
| C7H9ClN2 | |
| 25 g | |
| Hygroscopic | |
| Soluble in water. | |
| Cl.NC(=N)C1=CC=CC=C1 | |
| 156.61 | |
| 156.62 (Anhydrous) |
| 206752-36-5 | |
| water ≈10 to 14% | |
| MFCD00066285 | |
| 3594959 | |
| benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate | |
| LZCZIHQBSCVGRD-UHFFFAOYSA-N | |
| [amino(phenyl)methylidene]azanium | |
| 16219042 | |
| 98% |
Chemical Identifiers
| 206752-36-5 | |
| 156.61 | |
| LZCZIHQBSCVGRD-UHFFFAOYSA-N | |
| 16219042 |
| C7H9ClN2 | |
| MFCD00066285 | |
| benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate | |
| Cl.NC(=N)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 216-795-4
RTECSNumber : CV6260000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only