missing translation for 'onlineSavingsMsg'
Learn More

Benzamidine Hydrochloride 95.0+%, TCI America™

Catalog No. B33795G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B33795G Supplier TCI America Supplier No. B33795G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1670-14-0
Molecular Formula C7H9ClN2
Molecular Weight (g/mol) 156.613
MDL Number MFCD00013025
InChI Key LZCZIHQBSCVGRD-UHFFFAOYSA-N
Synonym benzamidine hydrochloride, benzimidamide hydrochloride, benzenecarboximidamide hydrochloride, benzenecarboximidamide, monohydrochloride, benzamidinium chloride, benzamidine hcl, benzamidine, hydrochloride, amidinobenzene hydrochloride, benzamidine, monohydrochloride, amidinobenzene hydrochloride 1:1
PubChem CID 80289
IUPAC Name benzenecarboximidamide;hydrochloride
SMILES C1=CC=C(C=C1)C(=N)N.Cl

Specifications

Melting Point 171°C
Color White
Quantity 5 g
Formula Weight 156.61
Percent Purity ≥95.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzamidine Hydrochloride [for Biochemical Research]
RTECSNumber CV6260000
TSCA Yes