missing translation for 'onlineSavingsMsg'
Learn More

Benzamidine hydrochloride, 99%

Catalog No. AC401790250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC401790250 25 g Glass bottle
AC401790050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC401790250 Supplier Thermo Scientific Chemicals Supplier No. 401790250
Only null left

CAS: 1670-14- | C7H9ClN2 | 156.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1670-14-
Molecular Formula C7H9ClN2
Molecular Weight (g/mol) 156.61
MDL Number MFCD00066285
InChI Key LZCZIHQBSCVGRD-UHFFFAOYSA-N
Synonym benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
PubChem CID 16219042
SMILES Cl.NC(=N)C1=CC=CC=C1

Specifications

Melting Point 167.0°C to 171.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C(NH2)NHC6H5·HCl
Quantity 25 g
Beilstein 09,28
Fieser 06,27
Solubility Information Solubility in water: soluble
IUPAC Name benzenecarboximidamide;hydrate;hydrochloride
Formula Weight 156.61
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Benzamidine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 216-795-4

RUO – Research Use Only