missing translation for 'onlineSavingsMsg'
Learn More

Benzamidine hydrochloride, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 401791000

Catalog No. AC401791000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Benzamidine hydrochloride
Authentic
Glass bottle
C(NH2)NHC6H5·HCl
100g
06,27
Solubility in water: soluble
NC(=[NH2+])C1=CC=CC=C1
121.16
156.61
1670-14-
98.5% min. (HPLC)
C7H9N2
MFCD00066285
09,28
benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
PXXJHWLDUBFPOL-UHFFFAOYSA-O
[amino(phenyl)methylidene]azanium
16219042
99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 216-795-4

SDS
Documents

Documents

Promotions

Promotions