missing translation for 'onlineSavingsMsg'
Learn More

Benzamide oxime, 97%, Thermo Scientific™

Catalog No. AC453280050
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453280050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC453280050 Supplier Thermo Scientific Chemicals Supplier No. 453280050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 613-92-3
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Molecular Formula C7H8N2O
Linear Formula C6H5C(NH2)=NOH
MDL Number MFCD00031485
Quantity 5g
Synonym benzamidoxime, benzamide oxime, n'-hydroxybenzimidamide, benzohydroxamamide, n-hydroxybenzamidine, phenylhydroxamidine, n-hydroxybenzenecarboximidamide, ccris 2953, n'-hydroxybenzamidine, benzamidoxim
InChI Key MXOQNVMDKHLYCZ-UHFFFAOYSA-N
SMILES N\C(=N/O)C1=CC=CC=C1
IUPAC Name N'-hydroxybenzenecarboximidamide
Molecular Weight (g/mol) 136.15
PubChem CID 7259353
ChEBI CHEBI:83354
Formula Weight 136.15
Percent Purity 97%
Chemical Name or Material Benzamide oxime
Show More Show Less
Hazard Category GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POIS
Hazard Statement GHS H Statement
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 210-361-8

RUO – Research Use Only