missing translation for 'onlineSavingsMsg'
Learn More

Benzamide, 98+%

Catalog No. p-7080797
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1050122 100 g
AAA1050136 500 g
AAA105010E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1050122 Supplier Thermo Scientific Chemicals Supplier No. A1050122

CAS: 55-21-0 | C7H7NO | 121.139 g/mol

Benzamide is utilized to study the mechanism of photocatalytic decomposition of aqueous solution of acetic acid, acetamide and acetonitrile in the presence of semiconductors. It is used as a nictoinamide-mimic PARP inhibitor and neuroprotectant. Further, it is used to develop a robust screening method to study biotransformations using (+)-gamma-lactamase enzyme. It is also employed in the determination of glycine. In addition to this, it is used as an intermediate in organic synthesis as well as in the production of pharmaceuticals and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzamide is utilized to study the mechanism of photocatalytic decomposition of aqueous solution of acetic acid, acetamide and acetonitrile in the presence of semiconductors. It is used as a nictoinamide-mimic PARP inhibitor and neuroprotectant. Further, it is used to develop a robust screening method to study biotransformations using (+)-gamma-lactamase enzyme. It is also employed in the determination of glycine. In addition to this, it is used as an intermediate in organic synthesis as well as in the production of pharmaceuticals and dyes.

Solubility
Soluble in water, methanol, ammonia, pyridine, carbon tetrachloride, carbon disulfide and ethanol. Slightly soluble in benzene and ethyl ether.

Notes
Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 55-21-0
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.139
MDL Number MFCD00007968
InChI Key KXDAEFPNCMNJSK-UHFFFAOYSA-N
Synonym benzoic acid amide, benzoylamide, phenylcarboxyamide, phenylcarboxamide, benzenecarboxamide, amid kyseliny benzoove, amid kyseliny benzoove czech, phenyl carboxyamide, ccris 4594, benzoic acid,amide
PubChem CID 2331
ChEBI CHEBI:28179
IUPAC Name benzamide
SMILES C1=CC=C(C=C1)C(=O)N

Specifications

Melting Point 125°C to 129°C
Density 1.341
Boiling Point 288°C
Flash Point 180°C (356°F)
Quantity 100 g
Beilstein 385876
Merck Index 14,1060
Solubility Information Soluble in water,methanol,ammonia,pyridine,carbon tetrachloride,carbon disulfide and ethanol. Slightly soluble in benzene and ethyl ether.
Formula Weight 121.14
Percent Purity ≥98%
Chemical Name or Material Benzamide
Show More Show Less
Hazard Category H302-H341
Hazard Statement GHS H Statement
H341-H302
Suspected of causing genetic defects.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
EINECSNumber 200-227-7
RTECSNumber CU8700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only