missing translation for 'onlineSavingsMsg'
Learn More

Benzalkonium chloride, 50% w/w aq. soln.

CAS: 63449-41-2 | C21H38ClN | 339.99 g/mol

Supplier:  Thermo Scientific Chemicals B207600F

 View more versions of this product

Catalog No. AAB207600F


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Benzalkonium chloride is a cationic surface active agent.It is used as a preservative in pharmaceutical products like eye, ear and nasal drops. It finds application in personal care products such as wet wipes, shampoos, hand sanitizers, deodorants and cosmetics. It is widely used in the chemical industry as a precipitant and emulsifier/de-emulsifier. It acts as corrosion inhibitor in the oil and gas industry pipeline by preventing the formation of sulfurous gases. It is also used in various non-consumer process like surgical disinfection.

Solubility
Miscible with water, methanol and acetone.

Notes
Incompatible with strong oxidizing agents, strong acids, strong bases and metals.
Specifications

Specifications

Benzalkonium chloride
0.99
C21H38ClN
2500 mL
4062599
benzyldimethyldecylammonium chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride 1:1, quaternary ammonium compounds, benzyl-c8-18-alkyldimethyl, chlorides, roccal, benzyl decyl dimethylazanium chloride, tret-o-lite xc 511, ccris 4586, benzyl decyl dimethylammonium chloride, c8-18-alkydimethylbenzyl ammonium chlorides
JBIROUFYLSSYDX-UHFFFAOYSA-M
benzyl-decyl-dimethylazanium;chloride
13762
63449-41-2
None
MFCD00137276
UN3265
14,1059
Miscible with water,methanol and acetone.
[Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
339.99
50% w/w aq. soln.

Chemical Identifiers

63449-41-2
339.99
JBIROUFYLSSYDX-UHFFFAOYSA-M
13762
[Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
C21H38ClN
MFCD00137276
benzyldimethyldecylammonium chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride 1:1, quaternary ammonium compounds, benzyl-c8-18-alkyldimethyl, chlorides, roccal, benzyl decyl dimethylazanium chloride, tret-o-lite xc 511, ccris 4586, benzyl decyl dimethylammonium chloride, c8-18-alkydimethylbenzyl ammonium chlorides
benzyl-decyl-dimethylazanium;chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 264-151-6

RTECSNumber : BO3151000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only