missing translation for 'onlineSavingsMsg'
Learn More

Benzaldehyde Diethyl Acetal 98.0+%, TCI America™

Catalog No. B189325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B189325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B189325G Supplier TCI America Supplier No. B189325G
Only null left

Chemical Identifiers

CAS 774-48-1
Molecular Formula C11H16O2
Molecular Weight (g/mol) 180.247
MDL Number MFCD00038525
InChI Key MAQMEXSLUSZDQM-UHFFFAOYSA-N
Synonym alpha,alpha-Diethoxytoluene
PubChem CID 69884
IUPAC Name diethoxymethylbenzene
SMILES CCOC(C1=CC=CC=C1)OCC

Specifications

Color Colorless
Boiling Point 222°C
Quantity 25 g
Formula Weight 180.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzaldehyde Diethyl Acetal
EINECSNumber (3)-1152
TSCA Yes