missing translation for 'onlineSavingsMsg'
Learn More

Bentonite, acid-leached, Thermo Scientific™

Catalog No. AC471231000
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC471231000 100 g
AC471230010 1 kg
AC471230050 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC471231000 Supplier Thermo Scientific Chemicals Supplier No. 471231000

Chemical Identifiers

CAS 70131-50-9

Specifications

Acidity 0.03 mg/KOH/g (Free Acidity)
Infrared Spectrum Conforms to structure
Packaging Plastic Bottle
Quantity 100 g
Titratable Acid ≤ 0.1 mg/KOH/g (Free Acidity)
Physical Form Granules
Chemical Name or Material Bentonite
ShelfLife 5 years
EINECSNumber 274-324-8
Recommended Storage Normal conditions