missing translation for 'onlineSavingsMsg'
Learn More

Benorylate, Thermo Scientific Chemicals

Catalog No. AC465960250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465960250 25 g
AC465960050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465960250 Supplier Thermo Scientific Chemicals Supplier No. 465960250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5003-48-5
Molecular Formula C17H15NO5
Molecular Weight (g/mol) 313.31
MDL Number MFCD00864257
InChI Key FEJKLNWAOXSSNR-UHFFFAOYSA-N
Synonym Benorilate
IUPAC Name 4-acetamidophenyl 2-(acetyloxy)benzoate
SMILES CC(=O)NC1=CC=C(OC(=O)C2=CC=CC=C2OC(C)=O)C=C1

Specifications

Alpha Vector BENORYLATE 001
Melting Point 177°C to 182°C
Color White
Loss on Drying ≤0.5 %
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 25 g
Formula Weight 313.31
Physical Form Crystalline Powder
Chemical Name or Material Benorylate
Hazard Category GHS07: Harmful
ShelfLife 5 years
storageNote1 Refrigerator +4°C

RUO – Research Use Only