missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Benorilate | |
| 180°C | |
| C17H15NO5 | |
| 25 g | |
| FEJKLNWAOXSSNR-UHFFFAOYSA-N | |
| 4-acetamidophenyl 2-(acetyloxy)benzoate | |
| 21102 | |
| ≥98.0% (HPLC,N) |
| 5003-48-5 | |
| White | |
| MFCD00864257 | |
| 4-Acetamidophenyl 2-Acetoxybenzoate, Aspirin Acetaminophen Ester | |
| CC(=O)NC1=CC=C(OC(=O)C2=CC=CC=C2OC(C)=O)C=C1 | |
| 313.31 | |
| 313.31 | |
| Crystalline Powder |
Chemical Identifiers
| 5003-48-5 | |
| 313.31 | |
| FEJKLNWAOXSSNR-UHFFFAOYSA-N | |
| 21102 | |
| CC(=O)NC1=CC=C(OC(=O)C2=CC=CC=C2OC(C)=O)C=C1 |
| C17H15NO5 | |
| MFCD00864257 | |
| 4-Acetamidophenyl 2-Acetoxybenzoate, Aspirin Acetaminophen Ester | |
| 4-acetamidophenyl 2-(acetyloxy)benzoate |
Safety and Handling
RTECSNumber : VO0720000
TSCA : No