missing translation for 'onlineSavingsMsg'
Learn More

Benorilate 98.0+%, TCI America™

Catalog No. B48275G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48275G 5 g
B482725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48275G Supplier TCI America Supplier No. B48275G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5003-48-5
Molecular Formula C17H15NO5
Molecular Weight (g/mol) 313.31
MDL Number MFCD00864257
InChI Key FEJKLNWAOXSSNR-UHFFFAOYSA-N
Synonym 4-Acetamidophenyl 2-Acetoxybenzoate, Aspirin Acetaminophen Ester
PubChem CID 21102
IUPAC Name 4-acetamidophenyl 2-(acetyloxy)benzoate
SMILES CC(=O)NC1=CC=C(OC(=O)C2=CC=CC=C2OC(C)=O)C=C1

Specifications

Melting Point 180°C
Color White
Quantity 5 g
Formula Weight 313.31
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Benorilate
RTECSNumber VO0720000
TSCA No