missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Benchtop Coolers

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.
Supplier: Thermo Scientific™ 51161300
Description
Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using Thermo Scientific™ Benchtop Coolers.
- Maintain freezer temperature during frost-free cycles, power failures and accidental door openings
- Holds 12, 20 or 32 tubes
- Inserts are included for 1.5 mL tubes to have the compatibility to hold 0.2-0.5 mL microcentrifuge tubes
- Printed grid on one-way keyed lid for proper orientation and sample identification
- Molded of impact-resistant polycarbonate with locking handle for long-term use
- Stackable to save space
Chemical Identifiers
| Polycarbonate | |
| Benchtop Cooler |
Specifications
| 190 mm | |
| 197 mm | |
| 140 mm | |
| 7.75 x 5.5 x 7.5 in. (197 x 140 x 190 mm) | |
| 3 x 4 | |
| Non-filled/clear lid | |
| Benchtop Cooler | |
| 0.2 to 0.5 mL | |
| 0°C |
| 7.5 in. | |
| 7.75 in. | |
| 5.5 in. | |
| 3x4 compartments, for 12-13mm dia.vials | |
| <1°C, 5.0 | |
| Polycarbonate | |
| Clear | |
| 12 Tubes (12 to 13 mm dia.) |