missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Benchtop Coolers

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.
Supplier: Thermo Scientific™ 51150012
Description
Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using Thermo Scientific™ Benchtop Coolers.
- Maintain freezer temperature during frost-free cycles, power failures and accidental door openings
- Holds 12, 20 or 32 tubes
- Inserts are included for 1.5 mL tubes to have the compatibility to hold 0.2-0.5 mL microcentrifuge tubes
- Printed grid on one-way keyed lid for proper orientation and sample identification
- Molded of impact-resistant polycarbonate with locking handle for long-term use
- Stackable to save space
Chemical Identifiers
| Polycarbonate | |
| Benchtop Cooler |
Specifications
| 125 mm | |
| 151 mm | |
| 108 mm | |
| 5.31 x 4.25 x 4.94 in. (151 x 108 x 125 mm) | |
| 3 x 4 | |
| Non-filled/clear lid | |
| Benchtop Cooler | |
| 0.2 to 0.5 mL | |
| -20°C |
| 4.94 in. | |
| 5.31 in. | |
| 4.25 in. | |
| 3x4 compartments, -20 deg. | |
| <-15°C, 1 | |
| Polycarbonate | |
| Blue, Clear | |
| 12 Tubes (0.2 to 0.5 mL) |