missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Benchtop Coolers
GREENER_CHOICE

Catalog No. 1535054
Change view
Click to view available options
Temperature Range:
-20°C
0°C
Array:
3 x 4
4 x 5
4 x 8
Holds:
12 Cryogenic Vials (0.2 to 1.5 mL)
12 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
12 Tubes (0.2 to 0.5 mL)
12 Tubes (12 to 13 mm dia.)
12 Tubes (16 to 17 mm dia.)
20 Tubes (0.2 to 0.5 mL)
32 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
32 Tubes (0.2 to 0.5 mL)
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range Array Holds
1535054 0°C 4 x 8 32 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
0974072 -20°C 4 x 5 20 Tubes (0.2 to 0.5 mL)
1535061 -20°C 3 x 4 12 Tubes (0.2 to 0.5 mL)
1535051 -20°C 4 x 8 32 Tubes (0.2 to 0.5 mL)
1535063 0°C 3 x 4 12 Tubes (12 to 13 mm dia.)
1535064 0°C 3 x 4 12 Tubes (16 to 17 mm dia.)
1535053 0°C 3 x 4 12 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
1535062 -20°C 3 x 4 12 Cryogenic Vials (0.2 to 1.5 mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 1535054 Supplier Thermo Scientific™ Supplier No. DS51160032

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using Thermo Scientific™ Benchtop Coolers.

  • Maintain freezer temperature during frost-free cycles, power failures and accidental door openings
  • Holds 12, 20 or 32 tubes
  • Inserts are included for 1.5 mL tubes to have the compatibility to hold 0.2-0.5 mL microcentrifuge tubes
  • Printed grid on one-way keyed lid for proper orientation and sample identification
  • Molded of impact-resistant polycarbonate with locking handle for long-term use
  • Stackable to save space

Specifications

Height (Metric) 146 mm
Height (English) 5.75 in.
Length (Metric) 243 mm
Length (English) 9.56 in.
Width (Metric) 157 mm
Width (English) 6.19 in.
Dimensions (L x W x H) 9.56 x 6.19 x 5.75 in. (243 x 157 x 146 mm)
Description 4x8 compartments, gel filled, 0 deg.
Array 4 x 8
Holds Temp., Hours <1°C, 5.0
Design Gel-filled/white lid
Material Polycarbonate
Product Type Benchtop Cooler
Color Blue, Clear
Capacity (Metric) 0.2 to 0.5 mL
Holds 32 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
Temperature Range 0°C
Show More Show Less