missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Benchtop Biohazard Disposal Can with Motion Sensor Lid

Catalog No. 16800565
Change view
Click to view available options
Capacity (Metric):
1.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
16800565 1.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16800565 Supplier Bel-Art™ Supplier No. H131941011
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Compact benchtop biohazard disposal can contains odors and is perfect for small items such as pipette tips and microcentrifuge tubes.

  • Automatic opening lid provides hands free use; closes automatically and securely for a more sanitary environment
  • Holds a 25 x 25 cm (10 x 10 in.) autoclavable biohazard disposal bag (F13166-000 or H13182-0000)
  • Includes a polypropylene liner with handle for easy emptying of contents
  • Aesthetic and durable; stainless steel body and chrome lid are resistant to bleach and easy to clean

Specifications

Capacity (Metric) 1.5 L
Certifications/Compliance CE and RoHS
Closure Type Chrome Lid
Diameter (Metric) Outer 12.4 cm
Diameter (English) Outer 4.8 in.
Capacity (English) 0.4 gal.
Color Silver
Height (Metric) 17 cm
Autoclavable Not Autoclavable
Includes Polypropylene liner with handle
Liner Material Polypropylene
For Use With (Application) Containing Odors; Sanitary Waste Disposal
For Use With (Equipment) 10 x 10 in. autoclavable biohazard disposal bag (F13166-000 or H13182-0000)
Height (English) 6.7 in.
Material Stainless Steel
Product Type Disposal Can
Show More Show Less