missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ BenchStable™ RPMI 1640
GREENER_CHOICE

Catalog No. p-7162107
Encompass
Change view
Click to view available options
Quantity:
500 mL
10 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A4192301 500 mL
A4192302 10 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A4192301 Supplier Gibco™ Supplier No. A4192301
Only null left

Gibco RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium) was originally developed to culture human leukemic cells in suspension and as a monolayer.

Gibco RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium) was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 medium has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of RPMI 1640 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

BenchStable RPMI 1640 Medium has been engineered for storage at room temperature while supporting cellular proliferation and maximum cell densities comparable to conventional RPMI 1640 Medium when supplemented with ≥10% fetal bovine serum (FBS).

This RPMI 1640 Medium is modified as follows:
• Optimized for room temperature stability
• With GlutaMAX, Phenol Red
• Without HEPES

Using BenchStable RPMI 1640 Medium
RPMI 1640 Medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 Medium contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. RPMI 1640 Medium uses a sodium bicarbonate buffer system (2.0 g/L) and so requires a 5–10% CO2 environment to maintain physiological pH. RPMI 1640 Medium with GlutaMAX supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. BenchStable RPMI 1640 Medium contains no proteins, lipids, or growth factors and so requires supplementation with ≥10% fetal bovine serum (FBS).

Specifications

Cell Line HeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium)
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
With Additives GlutaMAX, Phenol Red, Phenol Red
Without Additives No HEPES, No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 9001: 2015 standard
Product Line BenchStable
Quantity 500 mL
Shipping Condition Room Temperature
Green Features ACT Ecolabel (Silver)
Content And Storage Store at room temperature. Protect from light.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.