missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Infrared Micro-Sterilizer Accessories

Designed for use with Bacti-Loop™ micro sterilizer.

Supplier:  Fisherbrand™ B100003

 View more versions of this product

Catalog No. 22630003


Only null left

Description

Description

Slide Dryer Attachment for use with Fisher HealthCare™ Bacti-Loop™ Infrared Micro-Sterilizer

Order Info

Choice of accessory parts for use with Fisher HealthCare™ Bacti-Loop™ Infrared Micro-Sterilizer

Specifications

Chemical Identifiers

1.39 cm
6.5 in.
50/60 Hz
5 to 7 sec.
4 in.
Bacti-Loop Micro-Sterilizer
16.5 cm
Stainless Steel
120 V
10 cm

Specifications

6 in.
Slide dryer attachment, for 3 slides
1.39 cm
Bacti-Loop Micro-Sterilizer
16.5 cm
1500°F
2.9 lb.
120 V 50/60 Hz
Stainless Steel
120 V
10 cm
15 cm
0.55 in.
Ideal for platinum loops, needles and tube mouths
6.5 in.
Slide Dryer Attachment
815°C
1.4 kg
50/60 Hz
5 to 7 sec.
4 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions