LabExact Bench pads are designed with a complex proprietary fused 100% Polypropylene (PP) fibrous matrix which is a lightweight polyolefin and has an incredibly high chemical resistance. Bench pads are 290g/m2 basis weight sufficient to easily handle typical lab bench volume spills up to 600ml safely and quickly. Chemically resistant to most short term exposure of organic solvents as well as acids and alkalis. Lays flat as a bench pad with sufficient stiffness reducing bunch up seen with other competing brands. Soft enough to be used as a wipe. Binding dots accumulate fluids and migrate deeper into the pad allowing fast surface absorption and higher fluid hold saturation. 15 x 18", pack of 250.