missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Bench Mounted Reagent Poles

Catalog No. p-7114202
Change view
Click to view available options
Height (English):
12 in.
15 in.
18 in.
24 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English)
06000108 12 in.
06000109 15 in.
06000110 18 in.
06000111 24 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06000108
Only null left

Includes

Hardware

Fisherbrand™ Bench Mounted Reagent Poles are used to make stackable shelving on an island or peninsula. Poles can be stacked to make multiple tier shelving.

  • 1 in. Diameter painted steel pole is threaded at both ends
  • Bolts through the work surface for secure mounting
  • Fastener on the reagent shelf surface is recessed
  • Hardware is included for each installation
  • Poles are painted Fisherbrand White

Specifications

Product Type Reagent Pole
Height (Metric) 30.5 cm
Includes Installation Hardware
Diameter (English) 1 in.
Height (English) 12 in.
Depth (Metric) 2.5 cm
Color White
Width (Metric) 25 mm
Width (English) 1 in.
Material Steel