missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Bench Mounted Reagent Poles
Description
Includes
Hardware
- 1 in. Diameter painted steel pole is threaded at both ends
- Bolts through the work surface for secure mounting
- Fastener on the reagent shelf surface is recessed
- Hardware is included for each installation
- Poles are painted Fisherbrand White
Specifications
Specifications
| Product Type | Reagent Pole |
| Depth (Metric) | 2.5 cm |
| Diameter (English) | 1 in. |
| Color | White |
| Width (English) | 1 in. |
| Includes | Installation Hardware |
| Height (English) | 18 in. |
| Height (Metric) | 45.7 cm |
| Width (Metric) | 25 mm |
| Material | Steel |