missing translation for 'onlineSavingsMsg'
Learn More

Benazepril Hydrochloride 98.0+%, TCI America™

Catalog No. B36111G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36111G 1 g
B36115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36111G Supplier TCI America Supplier No. B36111G
Only null left

Chemical Identifiers

CAS 86541-74-4
Molecular Formula C24H29ClN2O5
Molecular Weight (g/mol) 460.96
MDL Number MFCD00895734
InChI Key VPSRQEHTHIMDQM-FKLPMGAJSA-N
Synonym benazepril hydrochloride, benazepril hcl, lotensin, cibacen, cibacene, briem, tensanil, zinadril, cibace, labopol
PubChem CID 5362123
ChEBI CHEBI:3012
IUPAC Name hydrogen 2-[(3S)-3-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl]acetic acid chloride
SMILES [H+].[Cl-].CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@H]1CCC2=CC=CC=C2N(CC(O)=O)C1=O

Specifications

Melting Point 183°C
Color White
Quantity 1 g
Formula Weight 460.96
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benazepril Hydrochloride
RTECSNumber CX7065000
TSCA No