missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Benazepril (hydrochloride)

Catalog No. AAH3798503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3798503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3798503 Supplier Thermo Scientific™ Supplier No. H37985MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 86541-74-4
Molecular Formula C24H29ClN2O5
Quantity 1 g
MDL Number MFCD00895734
InChI Key VPSRQEHTHIMDQM-FKLPMGAJSA-N
SMILES [H+].[Cl-].CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@H]1CCC2=CC=CC=C2N(CC(O)=O)C1=O
IUPAC Name hydrogen 2-[(3S)-3-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-1-yl]acetic acid chloride
Molecular Weight (g/mol) 460.96
Formula Weight 460.96
Physical Form Solid
ShelfLife 5 Years
EINECSNumber 630-414-2
RTECSNumber CX7065000