missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bullard™ Belts for EVA™ Powered-Air Purifying Respirators
Description
- Parts for EVA™ PAPR unit
- Choose from vinyl or foam material
- Can be paired with extenders
Specifications
Specifications
| Color | Black |
| Disposable | Reusable |
| Material | Vinyl |
| Product Type | Belt |
| For Use With (Equipment) | EVA™ PAPR Systems |
| Model | EVABELT2 |