missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ ProCulture™ Micro-Tube Homogenizer: Pestles and Tubes

Catalog No. p-4950196
Change view
Click to view available options
Packaging Type:
Bulk Pack
Individual Pack
Sample Volume (Metric):
0.5 mL
1.5 mL
Product Type:
Pestle
Pestle and Tube
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type Sample Volume (Metric) Product Type
03421220 Individual Pack 0.5 mL Pestle and Tube
03421221 Individual Pack 1.5 mL Pestle and Tube
03421227 Bulk Pack 1.5 mL Pestle and Tube
03421217 Individual Pack 1.5 mL Pestle
03421222 Bulk Pack 0.5 mL Pestle
03421223 Bulk Pack 1.5 mL Pestle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 03421220 Supplier Bel-Art™ Supplier No. F650000005
Only null left

Chemical and heat resistant, colorant-free

Matched pestle tips and micro tubes can be used either with homogenizer or with hand processing.

  • Tight tolerances, thorough homogenization and grinding of materials
  • Chemical and heat resistant, colorant-free to eliminate potential leaching of heavy metals into samples
  • Manufactured and packaged in class 10,000 cleanroom
  • Certified DNase, RNase and Pyrogen-free
  • Made of animal and phalate/phalate-ester free polypropylene
  • Available in individual or bulk packs
Pestles
  • Unique ribbed shaft provides precise finger control of homogenization for shear sensitive samples
  • Fit most motorized mixer adapters
Tubes
  • Tube bottom 25% thicker bottoms than industry standard
  • Frosted side and cap surfaces for labeling, positive seal snap-cap design and pierceable top

Specifications

Product Type Pestle and Tube
Voltage 0/1 V
Sample Volume (Metric) 0.5 mL
Material Polypropylene
Dnase Free Yes
For Use With (Equipment) Micro-Tube homogenizer system
Packaging Quantity 100/Pk.
Packaging Type Individual Pack
RNase Free Yes