Learn More
Bel-Art™ SP Scienceware™ 5mm O.D. Thin Walled Precision NMR Tubes
To maximize SNR, Precision NMR Tubes have minimal paramagnetic impurities that would impact shimming. Tight I.D. and O.D. tolerance as small as 0.0065mm accommodates Wilmad inserts
Supplier:  Bel-Art™ 665000575
 View more versions of this product
Catalog No. 16-800-432
Only null left
Explore available promotions
Alert:
Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.
Description
Description
To maximize SNR, Precision NMR Tubes have minimal paramagnetic impurities that would impact shimming. Tight I.D. and O.D. tolerance as small as 0.0065mm accommodates Wilmad inserts
Manufactured in a state-of-the-art ISO 9001:2008 USA facility using a unique precision shrinking and grinding process to shape the inner surface with maximized filling factor.
  • Inner surface is resistant to strong acid and base at ambient temperature
  • Can be operated safely at temperatures up to 230°C, and within a temperature step of 120°C
  • Rated as hydrolytic class 1
  • Made from borosilicate glass that meets the requirement of Type 1 Class A glass from ASTM E438
  • 100% inspection with multiple NIST traceable gauges and optical surface defect checks
Specifications
Specifications
NMR Tube
Glass (Type 1, Class A)
178 mm
4.2065±0.0065 mm
7 in.
528-PP-7-5
13 μm
25μm
4.9635±0.0065 mm
500MHz
0.38 mm
Product Suggestions
Product Suggestions