missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Waste Disposal Can with Lift-up Cover

Catalog No. 11394450
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
11394450 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
If using the container for biohazardous waste, then a biohazard label is required (not supplied).
Catalog No. 11394450 Supplier Bel-Art™ Supplier No. F131970000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

If using the container for biohazardous waste, then a biohazard label is required (not supplied).

Bright, red safety disposal container suitable for biological/biohazard materials, broken glass, recyclables or other waste

 

  • Sized for benchtop or floor placement
  • One piece hinged lid protects users from contact with contents; helps retain odors and contain airborne contaminants
  • Molded of polypropylene for long service life
  • Puncture-resistant; autoclavable at 121°C (250°F)
  • Fits 19 × 23 in. autoclave bags (sold separately)
  • Secured with an internal stainless steel wire bar

 

Specifications

Closure Type Hinged Top
Capacity (English) 5 gal.
Color Red
Dimensions (L x W x H) 9 x 12 x 11.5 in. (22.5 x 30.5 x 29 cm)
Disposable Single-use
Height (Metric) 29 cm
Length (Metric) 22.5 cm
Width (Metric) 30.5 cm
Autoclavable Autoclavable
For Use With (Application) Biological or biohazard materials, broken glass, recyclables or other waste
Width (English) 12 in.
Length (English) 9 in.
Height (English) 11.5 in.
Material Polypropylene
Product Type Safety Container
Show More Show Less