missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Vaccu-Pette 96-Well Format Pipetting Device, Autoclavable
Description
Requires
30mL or larger disposable sterile syringe
Aspirate or dispense 96 wells in less than 10 seconds, speeding up repetitive washing, dispensing or aspirating of wells by working simultaneously.
- For simultaneously changing media or the addition/removal of reagents from a 96 well plate
- Simply connect a disposable plastic syringe and withdraw the plunger to transfer liquid
- Made of transparent plastic material, it has vacuum outlet at one end, and a short length of 3.2mm I.D. tubing is supplied for syringe connection
- Autoclavable/Reusable model is molded of polycarbonate; must be sterilized by autoclaving at no more than 121°C for 20 minutes at 15 psi prior to use in order to avoid contamination
- Cannot be gas sterilized because of the possibility of residual trapped gases
- Requires a 30mL or larger disposable sterile syringe (not included)
Specifications
Specifications
| Adjustable Tip Spacing | No |
| Autoclavable | Autoclavable |
| Color | White |
| Disposable | Reusable |
| For Use With | 96-well Plates |
| Grip Type | Ergonomic |
| Material | Polycarbonate |
| Model | Vaccu-pette |
| Number of Channels | 96 |
| Product Line | SP Scienceware Vaccu-Pette |
| Show More |