Learn More
Bel-Art™ SP Scienceware™ Tubing Sinkers
Catalog No. 1387910
Change view
Click to view available options
Material:
PTFE
Catalog No. Material
1387910 PTFE
1 options
Catalog No. 1387910 Supplier Bel-Art™ Supplier No. F329030100
Get Up to $500 in Products with Purchase
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Excellent for use with pumps and dispensers, these TFE sinkers keep tubing at the bottom of a container
  • Sinkers have hose barb ends for simple tubing connection
  • Solvent resistant
  • Autoclavable
Specifications
Autoclavable Autoclavable
Material PTFE
Product Type Tubing Sinker