missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Super Glue

Catalog No. 1199924
Change view
Click to view available options
Volume (Metric):
3 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
1199924 3 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1199924 Supplier Bel-Art™ Supplier No. F240060000
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sets quickly to provide an amazingly strong repair or assembly joint

  • For use on metals, rubber, wood, most plastics or semi-porous materials
  • Dissimilar materials may be joined as well with a strong, dependable bond
  • Ethyl-cyanoacrylate adhesive, solvent-free and needs no catalyst or heat
  • Colorless and transparent when set

Specifications

Product Type Super Glue
Format Liquid
For Use With (Application) Used to join dissimilar materials like metals, rubber, wood, most plastics and semi-porous materials
For Use With (Equipment) Metal, Rubber, Wood, Plastics
Color Colorless
Material Ethyl-cyanoacrylate
Quantity 1 Ea.
Volume (English) 0.11 oz.
Volume (Metric) 3 g