missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Sterileware™ V-shaped Sampling Spatula, Sterile

Catalog No. 1435820
Change view
Click to view available options
Length (Metric):
22.9 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
1435820 22.9 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1435820 Supplier Bel-Art™ Supplier No. H36930 0000
Get Up to $500 in Products with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for every type of powdered or granulated materials. Spatulas are ideal for taking sterile samples during process quality inspections and for piercing plastic-lined paper bags or similar packages.

  • Time-honored V-shape makes transferring powdered, flaked, crystalline or granular media a breeze.
  • Long, 14 in. spatula (16-241-210) is perfect for dislodging caked or crystallized reagents from deep within tall bottles or vessels having a neck diameter as small as 0.75 in. (18 mm).
  • Sterileware (16-241-210, 14-385-20) available in 9 and 14 in. models.
  • High-impact polystyrene.
  • Individually packaged.
  • Sterilized by gamma irradiation.
  • Lot stamped, shipped with Certificate of Processing.
  • Nonsterile Model (03-421-149) 9 in. spatula.

Specifications

Disposable Single-use
Product Type Sterileware™ Sampling Spatula
For Use With (Equipment) Containers, bags and drums
For Use With (Application) Ideal for every type of powdered or granulated material, perfect for taking sterile samples during process quality inspections and for piercing Plastic-lined Paper bags or similar package
Length (English) 9 in.
Width (English) 0.75 in.
Width (Metric) 1.9 cm
Dimensions (L x W) 9 x 0.75 in. (22.9 x 1.9 cm)
Includes 6 in. L V-shaped channel
Length (English) Handle 2.99 in.
Length (Metric) Handle 7.6 cm
Material Polystyrene
Length (Metric) 22.9 cm
Sterility Sterile
Autoclavable Not Autoclavable
Shape Pointed End
Show More Show Less