Learn More
Bel-Art™ Sterileware™ Economy Sample Spoons
Molded high-impact polystyrene for durability and stiffness. Ideal for retrieving small, specific volume samples of powders or liquids.
$58.30 - $484.47
| Catalog Number | Mfr. No. | Capacity (Metric) | Sterility | Quantity | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Sterility | Quantity | Price | Quantity | |||
|
03-410-637
|
Bel-Art™
F367400022 |
2.5 mL | Non-sterile | 12/Pk. |
Pack of 12 Each for $58.30
|
|
|||
|
03-410-634
|
Bel-Art™
F367400004 |
8 mL | Sterile | 100/Pk. |
Pack of 100 Each for $484.47
|
|
|||
|
03-410-631
|
Bel-Art™
F367400001 |
1.25 mL | Sterile | 100/Pk. |
Pack of 100 Each for $430.67
|
|
|||
Description
- Molded of durable, white high-impact polystyrene for durability and stiffness when sampling packed powders
- Flat-bottom shape permits use as a weighing boat
- For one-time use
Order Info
100 individually sterile wrapped spoons or 12 nonsterile spoons per pack
Specifications
| Not Autoclavable | |
| Spoon |