missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Vaccu-Pette 96-Well Format Pipetting Device

Catalog No. 1371216
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1371216 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For single use only: Cannot be autoclaved or gas sterilized
Catalog No. 1371216 Supplier Bel-Art™ Supplier No. F378760000
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For single use only: Cannot be autoclaved or gas sterilized

Aspirate or dispense 96 wells in less than 10 seconds, speeding up repetitive washing or dispensing

  •  For simultaneously changing media or addition/removal of reagents from 96-well plate
  • Connect disposable plastic syringe and withdraw plunger to transfer liquid
  • Made of transparent plastic material
  • With vacuum outlet at one end, and short tube of 3.2mm I.D. supplied for syringe connection
  • Sterile/Disposable model molded of clear, disposable plastic
  • Requires 30mL or larger disposable sterile syringe

Specifications

Description 96 wells; L x W x H: 12.7 x 8.6 x 3.7cm
Material Plastic
Sterility Sterile
Quantity Each
Color Clear
Volume (Metric) 30 mL
Length (Metric) 12.7 cm
Type Pipetting Device
Disposable Reusable