Learn More
Bel Art™ ProCulture™ Stak-A-Tray™ System
Maximize incubator, cold room and bench space.
Specifications
Autoclavable Autoclavable
Color Silver
Material Stainless Steel
Positions 12
Stackable Yes
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Holds Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Holds Price Quantity & Availability  
079104A
View Documents Promotion Details
Bel Art™
H186100470
Small Tray Holder Sample Containers N/A
Only null left
079106
View Documents Promotion Details
Bel Art™
H186101440
Large Tray Holder Sample Containers N/A
Only null left
Description
Description
  • Keeps trays, flasks and dishes level and stable for even settling of tissue culture cells
  • Racks interlock when stacked, to a maximum of 5 racks high (allows up to 20 levels of trays)
  • Each rack tier holds one large tray (Mfr. No. 18610-1440), 35.6 x 35.6cm, or two smaller trays (Mfr. No. 18610-0470), 17.8 x 35.6cm
  • Perforated trays allow air, gas and water vapor circulation
  • Racks and trays sold separately
  • Stainless-steel storage system is autoclavable
Rack, 2-Tier (Mfr. No. 18610-0420)
  • 5cm clearance between trays, ideal for storing larger culture flasks
Rack, 4-Tier (Mfr. No. 18610-0440)
  • 2.4cm clearance between trays, for storing Petri dishes or smaller culture flasks
Specifications
Specifications
Autoclavable
Stainless Steel
Yes
12 x 1
Silver
12
None