Learn More
Bel-Art™ Spinfin™ Magnetic Stirring Bar
Agitate liquids in round bottom vessels like test tubes, cylinders and flasks
$29.10
Specifications
Autoclavable Autoclavable
Certifications/Compliance FDA, USP Class VI, ISO 9001:2008, SGS
Color White
Coating PTFE, FDA-grade, USP Class VI
Material Alnico Magnet
View More Specs
Products 1
Catalog Number Mfr. No. Diameter (Metric) Diameter (English) Height (Metric) Height (English) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Diameter (English) Height (Metric) Height (English) Price Quantity & Availability  
14512166
View Documents Promotion Details
Bel-Art™
F371250014
7.9 mm 0.31 in. 9.5mm 0.37 in.
Each for $29.10
Only null left
Description
Description
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE low friction coating
  • Does not wobble in vessel
  • Autoclave safe
Certifications
ISO 9001:2008, SGS certification.
Specifications
Specifications
Autoclavable
White
Alnico Magnet
Cross
FDA, USP Class VI, ISO 9001:2008, SGS
PTFE, FDA-grade, USP Class VI
50
Magnetic Stir Bar