missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Spinbar™ Magnetic Stirring Bar Restrainer

Catalog No. 03421110
Change view
Click to view available options
Length (Metric):
8 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
03421110 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03421110 Supplier Bel-Art™ Supplier No. F377861000
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fits ergonomically in one hand. Bel Art™ Spinbar™ Magnetic Stirring Bar Restrainers retains and secures the stirring bar in position.

  • Eliminate hazard caused with stir bar splashes when decanting mixing vessels
  • After the vessel is drained, the confined stirring bar is securely retrieved from the vacant container without contaminating the solution
  • Fabricated with simple-to-clean durable polystyrene
  • Houses strong magnet that restrains magnetic stirring bars up to 80 mm (3 in.) long

Specifications

Material Polyethylene, Alnico Magnet
Dimensions (L x W x H) 51 x 9.5 mm
Shape Disc
Diameter (Metric) 51 mm
Quantity 1
Length (Metric) 8 cm
Type Spinbar™ Magnetic Stirring Bar Restrainer
For Use With (Equipment) Stirring bars
For Use With (Application) Eliminate hazard caused with stir bar splashes when decanting mixing vessels, capture bars in containers and restrains bars up to 3 in. long
Length (English) 3 in.
Description Fabricated with simple-to-clean durable polypropylene
Show More Show Less