missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Secador™ Gas-Purge Desiccator Cabinet

Catalog No. p-3493943
Change view
Click to view available options
Height (Metric):
21.3 cm
31.4 cm
41.7 cm
51.8 cm
Color:
Amber
Clear
No. of Shelves:
1
1 Fixed Vented Adjustable
2
2 vented adjustable shelves
3
3 perforated removable shelves
Model:
1.0
2.0
3.0
4.0
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric) Color No. of Shelves Model
08647663 31.4 cm Clear 2 vented adjustable shelves 2.0
08647666 41.7 cm Clear 3 perforated removable shelves 3.0
14809178 51.8 cm Clear 2 vented adjustable shelves 4.0
14809177 21.3 cm Clear 1 Fixed Vented Adjustable 1.0
0181620 51.8 cm Clear 3 4.0
08647721 31.4 cm Amber 2 2.0
08647722 41.7 cm Amber 3 3.0
08647720 21.3 cm Amber 1 1.0
08647723 51.8 cm Amber 3 4.0
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 08647663 Supplier Bel Art™ Supplier No. F420721002
Only null left

Available in a wide range of sizes for gas-purged desiccation.

Available in a wide range of sizes for gas-purged desiccation.

  • Create a controlled environment with dry nitrogen desiccation or other gases of choice; Dust-free and desirable for many applications including clean room environments
  • Rear mounted gas port valves accept 0.25 in. tubing and can be closed to use standard desiccants
  • The attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products; Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing, and chemical attack
  • Amber color models reduce visible light penetration by over 50% for optimal storage light-sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf
  • Models 1.0, 2.0, 3.0 and 4.0: Vertical units have a compact 16.3 L x 13.4 in.W footprint for space-saving stacking up to 3 units high and side swinging door that opens to the left; 4.0 horizontal has a wider footprint, lower height and pull down oven style door
  • U.S. Patents: 6,834,920; 7,318,630 and D474,626

Dimensions

  • Secador 1.0: 0.74 cu. ft. internal volume, 16.3 L x 13.4 W x 8.4 in. H
  • Secador 2.0: 1.16 cu. ft. internal volume, 16.3 L x 13.4 W x 12.4 in. H
  • Secador 3.0: 1.55 cu. ft. internal volume, 16.3 L x 13.4 W x 16.4 in.H
  • Secador 4.0: 1.87 cu. ft. internal volume, 16.3 L x 13.4 W x 20.4 in. H

Order Info

Use with desiccant cartridges Mfr. Nos. 420450250, 420450200, 420480100. Replacement shelves available: 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves; Replacement Shelf for Horizontal 4.0 Cabinets, Mfr. No. 420740001 or Replacement Shelf for Vertical 1.0, 2.0, 3.0 or 4.0 Cabinets, Mfr. No. 420741001.

Specifications

Autoclavable Not Autoclavable
Color Clear
Cabinet Style Vertical
Material Copolyester
Product Type Desiccator
Door Style Left-hinged and gasketed
For Use With (Application) Create a controlled environment with dry nitrogen desiccation or other gases of choice, dust-free and desirable for many applications including clean room environments
Includes Rear mounted gas port valves, carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
Model 2.0
No. of Shelves 2 vented adjustable shelves
Resistance Chemicals, stains and fine cracks
Volume (English) 1.16 cu. ft.
Dimensions (L x W) 16.3 x 13.4 in. (41.4 x 34.1 cm)
Length (English) 16.3 in.
Length (Metric) 41.4 cm
Width (English) 13.4 in.
Width (English) Door 8.9 in.
Width (Metric) 34.1 cm
Width (Metric) Door 22.5 cm
Height (English) 12.4 in.
Height (English) Opening 8.4 in.
Height (Metric) 31.4 cm
Depth (English) 16.3 in.
Depth (Metric) 41.4 cm
Show More Show Less