missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel Art™ Secador™ Gas-Purge Desiccator Cabinet
Description
Available in a wide range of sizes for gas-purged desiccation.
- Create a controlled environment with dry nitrogen desiccation or other gases of choice; Dust-free and desirable for many applications including clean room environments
- Rear mounted gas port valves accept 0.25 in. tubing and can be closed to use standard desiccants
- The attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products; Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing, and chemical attack
- Amber color models reduce visible light penetration by over 50% for optimal storage light-sensitive materials like reagents, analytical standards and investigational compounds
- Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
- Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf
- Models 1.0, 2.0, 3.0 and 4.0: Vertical units have a compact 16.3 L x 13.4 in.W footprint for space-saving stacking up to 3 units high and side swinging door that opens to the left; 4.0 horizontal has a wider footprint, lower height and pull down oven style door
- U.S. Patents: 6,834,920; 7,318,630 and D474,626
Dimensions
- Secador 1.0: 0.74 cu. ft. internal volume, 16.3 L x 13.4 W x 8.4 in. H
- Secador 2.0: 1.16 cu. ft. internal volume, 16.3 L x 13.4 W x 12.4 in. H
- Secador 3.0: 1.55 cu. ft. internal volume, 16.3 L x 13.4 W x 16.4 in.H
- Secador 4.0: 1.87 cu. ft. internal volume, 16.3 L x 13.4 W x 20.4 in. H
Order Info
Use with desiccant cartridges Mfr. Nos. 420450250, 420450200, 420480100. Replacement shelves available: 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves; Replacement Shelf for Horizontal 4.0 Cabinets, Mfr. No. 420740001 or Replacement Shelf for Vertical 1.0, 2.0, 3.0 or 4.0 Cabinets, Mfr. No. 420741001.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Color | Amber |
| Cabinet Style | Vertical |
| Material | Copolyester |
| Product Type | Desiccator |
| Door Style | Left-hinged and gasketed |
| For Use With (Application) | Create a controlled environment with dry nitrogen desiccation or other gases of choice, dust-free and desirable for many applications including clean room environments |
| Includes | Rear mounted gas port valves, carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer |
| Model | 4.0 |
| No. of Shelves | 3 |
| Show More |