Learn More
Bel-Art™ Secador™ Desiccator Replacement Shelves
Catalog No. 0181619
Change view
Click to view available options
Product Type:
Desiccator Shelf (Vertical)
Catalog No. Product Type
0181619 Desiccator Shelf (Vertical)
1 options
Catalog No. 0181619 Supplier Bel-Art™ Supplier No. F420741001
Get Up to $500 in Products with Purchase
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Convert 4.0 units from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves.
  • Fit auto and nonelectric models
  • Durable, clear polystyrene construction
  • Perforated to faciilitate air flow
Specifications
Diameter (Metric) Outer 51.8 cm
Material Polystyrene
For Use With (Application) Convert 4.0 units from horizontal to vertical desiccator type
Product Type Desiccator Shelf (Vertical)