missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Secador™ Auto Desiccator Cabinet, Vertical Model

Catalog No. 08647858
Change view
Click to view available options
Color:
Amber
Clear
Cabinet Style:
Vertical
Vertical (Tinted)
Voltage:
120 V
Volume (English):
1.16 cu. ft.
1.55 cu. ft.
1.87 cu. ft.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Cabinet Style Voltage Volume (English)
08647858 Amber Vertical (Tinted) 120 V 1.87 cu. ft.
0864752 Clear Vertical 120 V 1.87 cu. ft.
08647655 Clear Vertical 120 V 1.55 cu. ft.
08647656 Clear Vertical 120 V 1.16 cu. ft.
08647852 Amber Vertical (Tinted) 120 V 1.16 cu. ft.
08647855 Amber Vertical (Tinted) 120 V 1.55 cu. ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 08647858 Supplier Bel Art™ Supplier No. F420741118
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A UV light blocking, dry environment with automatic electronic dehumidification.

A UV light blocking, dry environment with automatic electronic dehumidification.

  • Attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products.
  • Patented electronic dehumidification system reduces humidity and circulates dry air automatically, freeing users from monitoring and changing desiccants.
  • Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack.
  • Amber color models reduce visible light penetration by over 50% for storing light sensitive materials like reagents, analytical standards and investigational compounds.
  • Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals.
  • Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf.
  • 4.0 horizontal has a wider footprint, lower height and pull down oven style door.
  • U.S. Patents: 6,772,534; 6,834,920; 7,114,266; 7,318,630 and D474,626.
  • 4.0 units can be converted from horizontal to vertical configuration or vice versa by turning the unit and ordering the appropriate number of shelves (see Ordering Info).
  • Secador 2.0: 1.16 cu. ft. internal volume, 16.3 L x 13.4 W x 12.4 in. H
  • Secador 3.0: 1.55 cu. ft. internal volume, 16.3 L x 13.4 W x 16.4 in. H
  • Secador 4.0: 1.87 cu. ft. internal volume, 16.3 L x 13.4 W x 20.4 in. H

Order Info

Replacement shelves are available.

Specifications

Autoclavable Not Autoclavable
Color Amber
Cabinet Style Vertical (Tinted)
Material Copolyester
Product Type Desiccator
Door Style Side-swinging
For Use With (Application) Easy and reliable storage of moisture and light sensitive products, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant
Includes Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
Model 4.0
No. of Shelves 3
Resistance Chemicals, stains and fine cracks
Shelf Type Removable Perforated
Temperature Range (English) 41°F to 104°F
Temperature Range (Metric) 5°C to 40°C
Volume (English) 1.87 cu. ft.
Dimensions (L x W) 16.3 x 13.4 in. (41.4 x 34.1 cm)
Length (English) 16.3 in.
Length (Metric) 41.4 cm
Width (English) 13.4 in.
Width (Metric) 34.1 cm
Height (English) 20.4 in.
Height (Metric) 51.8 cm
Depth (English) 16.3 in.
Depth (Metric) 41.4 cm
Voltage 120 V
Show More Show Less