missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel Art™ Secador™ Auto Desiccator Cabinet, 4.0 Horizontal Model
A UV light blocking, dry environment with automatic electronic dehumidification.
$2,401.27
Specifications
| Autoclavable | Not Autoclavable |
|---|---|
| Cabinet Style | Horizontal |
| Material | Copolyester |
| Product Type | Desiccator |
| Door Style | Bottom-hinged, pull down oven style |
| Catalog Number | Mfr. No. | Color | Shelf Type | Voltage | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Shelf Type | Voltage | Price | Quantity & Availability | |||
|
0864756
|
Bel Art™
F420740115 |
Clear | Vented Adjustable | 120 V |
Each for $2,401.27
|
|
|||
Description
- Attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products.
- Patented electronic dehumidification system reduces humidity and circulates dry air automatically, freeing users from monitoring and changing desiccants.
- Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack.
- Amber color models reduce visible light penetration by over 50% for storing light sensitive materials like reagents, analytical standards and investigational compounds.
- Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals.
- Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf.
- 4.0 horizontal has a wider footprint, lower height and pull down oven style door.
- U.S. Patents: 6,772,534; 6,834,920; 7,114,266; 7,318,630 and D474,626.
- 4.0 units can be converted from horizontal to vertical configuration or vice versa by turning the unit and ordering the appropriate number of shelves (see Ordering Info).
- Secador 2.0: 1.16 cu. ft. internal volume, 16.3 L x 13.4 W x 12.4 in. H
- Secador 3.0: 1.55 cu. ft. internal volume, 16.3 L x 13.4 W x 16.4 in. H
- Secador 4.0: 1.87 cu. ft. internal volume, 16.3 L x 13.4 W x 20.4 in. H
Order Info
Replacement shelves are available: Replacement Shelf for Horizontal 4.0 Cabinets, Mfr. No. 420740001
Specifications
| Not Autoclavable | |
| Copolyester | |
| Bottom-hinged, pull down oven style | |
| Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer | |
| 2 | |
| 41°F to 104°F | |
| 20.4 in. | |
| 13.4 in. | |
| 16.3 in. |
| Horizontal | |
| Desiccator | |
| Easy and reliable storage of moisture and light sensitive products, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant | |
| 4.0 | |
| Chemicals, stains and fine cracks | |
| 5°C to 40°C | |
| 51.8 cm | |
| 34.1 cm | |
| 41.4 cm |