Learn More
Bel-Art™ Replacement Stopcocks
Catalog No. 22261910
Change view
Click to view available options
Plug Bore:
0.24 in. x 0.38 in. O.D.
Catalog No. Plug Bore
22261910 0.24 in. x 0.38 in. O.D.
1 options
Catalog No. 22261910 Supplier Bel-Art™ Supplier No. F420400000
Get Up to $500 in Products with Purchase
View all promotions
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Polypropylene body has Teflon™ PTFE plug, washer and bright red handle
  • Three-way plug for vacuum draw, shut-off and vacuum release
  • Taper fits on desiccator porte
Use on Desiccator Models: 08-594-15A, 08-594-16A, 08-594-15B, 08-594-16B, 08-594-15C, 08-594-16C, 01-817-50
Specifications
Type 3-way Stopcock
Plug Bore 0.24 in. x 0.38 in. O.D.
Material Polypropylene
For Use With (Application) Vacuum Desiccator Cabinets
For Use With (Equipment) Scienceware™ desiccators, space saver and Techni-Dome™ 360 models
Includes Washer, high visibility bright red handle with nut
Plug Material PTFE
Outlet Port Male Slip
Connectors 3-way
Teflon™ is a registered trademark of the Chemours Company FC LLC used under license by Bel-Arts Products, Inc.